Owl™ Dual-Gel Vertical Electrophoresis Systems
Thermo Scientific™

Owl™ Dual-Gel Vertical Electrophoresis Systems

Quickly and easily cast and run one or two gels in the same device with these Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric) BufferIncludes
P8DS-1300 mLComplete System plus Caster
P10DS-11250 mLComplete System plus Caster
P10DS-21250 mLComplete System
P10DS-31250 mLApparatus Only
P8DS-2300 mLComplete System
P8DS-3300 mLApparatus Only
Catalog number P8DS-1
Price (USD)
1,629.00
Online exclusive
1,810.00
Save 181.00
Each
Volume (Metric) Buffer:
300 mL
Includes:
Complete System plus Caster
Price (USD)
1,629.00
Online exclusive
1,810.00
Save 181.00
Each

Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.

Simple, Six-Step Use:

  1. Secure the glass cassettes
  2. Pour the agarose
  3. Plug in the specially designed trough
  4. Cast polyacrylamide solution in the cassettes
  5. Load samples
  6. Run the gel

Features:

  • Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
  • Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
  • Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
  • Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free

Recommended for:

  • SDS electrophoresis
  • The second dimension of two-dimensional (2-D) electrophoresis
  • Agarose electrophoresis
  • Native electrophoresis
Specifications
Gel Dimensions10 x 8-10 cm
Height (English)5.5 in.
IncludesComplete System plus Caster
Length (English)6.7 in.
Length (Metric) Gel8 to 10 cm
ModelP8DS
Volume (Metric) Buffer300 mL
Width (English)5.1 in.
Width (Metric) Gel10 cm
Product LineOwl
TypeDual-Gel Vertical Electrophoresis Complete System with Caster
Unit SizeEach

Frequently asked questions (FAQs)

Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.