Accessories for TSG Series Countertop Refrigerators
Product Image
Thermo Scientific™
Accessories for TSG Series Countertop Refrigerators
Innovative refrigeration solutions for healthcare and research needs
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
PA02Pyxis Lock Adapter
OF02Omnicell Lock Adapter
RB02TS
also known as RB02
Roller Base
Catalog number PA02
Price (USD)
197.10
Online Exclusive
219.00
Save 21.90 (10%)
Each
Add to cart
Type:
Pyxis Lock Adapter
Price (USD)
197.10
Online Exclusive
219.00
Save 21.90 (10%)
Each
Add to cart
Thermo Scientific™ TSG Series Countertop Refrigerators have several accessory options available. E-lock adapter kits are available for Pyxis™ and Omnicell™ systems, and a 8.5 in. rollerbase makes moving the TSG205SA from room-to-room easy and convenient.
  • OF02 is compatible with Omnicellock™ systems
  • PA02 is compatible with Pyxis™lock systems
  • RB02 is an 8.5" roller base for the TSG205SA
Specifications
Access SecurityPyxis Lock Adapter
For Use With (Equipment)TSG Series Countertop Refrigerator
TypePyxis Lock Adapter
StyleCountertop
Unit SizeEach