Hyaluronic acid Polyclonal Antibody - FAQs
Hyaluronic acid Polyclonal Antibody - FAQs
View additional product information for Hyaluronic acid Polyclonal Antibody - FAQs (PA185561)
2 product FAQs found
How should I prepare my plates for a direct ELISA using the Hyaluronic acid Polyclonal Antibody (Cat. No. PA1-85561)?
For direct ELISA, we recommend coating wells with 1% glutaraldehyde for 5 min at room temperature, followed by 3X PBS wash steps before coating with the Hyaluronic acid Polyclonal Antibody (Cat. No. PA1-85561).
What is the molecular weight of the Hyaluronic acid Polyclonal Antibody (Cat. No. PA1-85561)?
The average molecular weight for the Hyaluronic acid Polyclonal Antibody (Cat. No. PA1-85561) is 150 kDa.