SSTR2 Polyclonal Antibody - FAQs
SSTR2 Polyclonal Antibody - FAQs
View additional product information for SSTR2 Polyclonal Antibody - FAQs (PA316736)
2 product FAQs found
The antibody I purchased from you says ''concentration not determined'' on the product label and on the product page. How do I determine the concentration?
Some antibodies are provided in non-purified formats, such as ascites fluid, tissue culture supernatant, or whole serum. These formats contain multiple proteins, which can interfere with accurate protein quantification. Traditional methods like A280 measurement or protein assays such as BCA quantify total protein but do not provide a specific concentration for the antibody in question.
Literature, such as "Antibodies" by Harlow & Lane (1988) (https://antibodiesmanual.org/index.php) (Table 8.2 - Storing and Purifying Antibodies), suggests approximate concentrations of antibodies in these solutions, as shown below. However, please note that these are rough estimates, and actual concentrations can vary based on several factors.
Serum
Total antibody concentration: 10 mg/mL
Specific antibody concentration: 0.1 to 1 mg/mL (a maximum of 10% of the antibody population is expected to be the specific antibody of interest)
Tissue Culture Supernatant (10% FBS)
Total antibody concentration: 1 mg/mL
Specific antibody concentration: 0.05 mg/mL
Tissue Culture Supernatant (serum-free)
Total antibody concentration: 0.01 to 0.05 mg/mL
Specific antibody concentration: 0.01 to 0.05 mg/mL
Ascites
Total antibody concentration: 1 to 10 mg/mL
Specific antibody concentration: 0.9 to 9 mg/mL
Do you offer SSTR2 antibodies for western blot detection?
We offer 5 SSTR2 polyclonal antibodies that target human and have been verified for western blot detection (https://www.thermofisher.com/antibody/primary/target/sstr2/filter/proteintarget/SSTR2/application/Western%20Blot/species/Human). Their tested dilutions for western blot detection are listed below.
Catalog Number and related western blot dilution information:
PA3-109    1:250
711091     1-2 µg/mL
PA3-16736   1:100-1:1000
PA5-85431   1:500-1:3000
BS-1138R   Assay Dependent
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.