SSTR2 Polyclonal Antibody - FAQs
SSTR2 Polyclonal Antibody - FAQs
View additional product information for SSTR2 Polyclonal Antibody - FAQs (PA534221)
1 product FAQs found
Can I use SSTR2 Antibody (Cat. No. PA5-34221) for western blot detection?
Unfortunately,the SSTR2 Antibody (Cat. No. PA5-34221) has not been internally verified for western blot detection, and therefore we cannot guarantee that it will work for this application.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.