Do you offer SLC22A8 Polyclonal Antibody (Cat. No. PA5-60639)?
SLC22A8 Polyclonal Antibody (Cat. No. PA5-60639) has been discontinued. As an alternative, you may use SLC22A8 Polyclonal Antibody (Cat. No. PA5-102699).
Search
View additional product information for SLC22A8 Polyclonal Antibody - FAQs (PA560639)
1 product FAQs found