IL-2 Polyclonal Antibody - FAQs
IL-2 Polyclonal Antibody - FAQs
View additional product information for IL-2 Polyclonal Antibody - FAQs (PA585127)
1 product FAQs found
Can you provide the exact immunogen sequence for IL-2 Polyclonal Antibody (Cat. No. PA5-85127)?
The exact immunogen sequence for IL-2 Polyclonal Antibody (Cat. No. PA5-85127) is proprietary. However, we can divulge that the immunogen is a recombinant protein encompassing a sequence within the center region of human IL-2.