Phospho-p53 (Thr18) Polyclonal Antibody - FAQs
Phospho-p53 (Thr18) Polyclonal Antibody - FAQs
View additional product information for Phospho-p53 (Thr18) Polyclonal Antibody - FAQs (PA512660)
1 product FAQs found
What is the recommended blocking buffer when using the Phospho-p53 (Thr18) Polyclonal Antibody (Cat. No. PA5-12660) for the detection of phosphorylation of a protein?
We recommend using BSA as the blocking buffer when using the Phospho-p53 (Thr18) Polyclonal Antibody (Cat. No. PA5-12660) for the detection of protein phosphorylation.