What is the recommended dilution for the FOXG1 Polyclonal Antibody (Cat. No. PA526794) in immunocytochemistry/immunofluorescence (ICC/IF) applications?
We recommend using a dilution range of 1:10-1:50 when using the FOXG1 Polyclonal Antibody (Cat. No. PA526794) in immunocytochemistry/immunofluorescence (ICC/IF) applications.