Is the FOXG1 Polyclonal Antibody (Cat. No. PA526794) validated for immunocytochemistry/immunofluorescence (ICC/IF) applications?
Yes, the FOXG1 Polyclonal Antibody (Cat. No. PA526794) has been validated for ICC/IF applications.
Search
View additional product information for LMBR1L Polyclonal Antibody - FAQs (PA526795)
1 product FAQs found