Does the Transferrin Receptor Polyclonal Antibody (Cat. No. PA527739) have cross-reactivity to chickens or avians, besides humans and mouse?
There is a 58.8% identity in 160 residues overlap with this chicken sequence in the UniProt Database. Unfortunately, we don’t have experimental data to show whether or not 58.8% homology can sufficiently show cross-reactivity with chicken or avians.