PGP9.5 Polyclonal Antibody - FAQs
PGP9.5 Polyclonal Antibody - FAQs
View additional product information for PGP9.5 Polyclonal Antibody - FAQs (PA529012)
1 product FAQs found
Do you offer an alternative for PGP9.5 Polyclonal Antibody (Cat. No. PA5-16825)?
A suggested replacement for PGP9.5 Polyclonal Antibody (Cat. No. PA5-16825), 500 µL, is PGP9.5 Polyclonal Antibody (Cat. No. PA5-29012), 100 µL. The main difference between Cat. No. PA5-16825 and Cat. No. PA5-29012, is that Cat. No. PA5-16825 immunogen is a recombinant full-length human PGP9.5 protein, while the alternative Cat. No. PA5-29012 immunogen is a synthetic peptide corresponding to a region within amino acids 137 and 223 of the human PGP9.5 protein.