LGR5 Polyclonal Antibody - FAQs
LGR5 Polyclonal Antibody - FAQs
View additional product information for LGR5 Polyclonal Antibody - FAQs (PA533728)
1 product FAQs found
What was the pH of the buffer used for validating the LGR5 Polyclonal Antibody (Cat. No. PA5-33728)?
The pH of the buffer used for validating the ASCL2 Polyclonal Antibody (Cat. No. PA5-33728) was pH 9-9.5.