Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
A suggested positive control for Western blot is mouse heart cells; suggested positive control for IHC is human breast carcinoma.
Cofilin is a widely distributed intracellular actin-modulating protein that binds and depolymerizes filamentous F-actin and inhibits the polymerization of monomeric G-actin in a pH-dependent manner. It is involved in the translocation of actin-cofilin complex from cytoplasm to nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 18 kDa phosphoprotein; CFL-1; cofilin 1 (non-muscle); cofilin 2 (muscle); Cofilin, muscle isoform; Cofilin, non-muscle isoform; Cofilin-1; Cofilin-2; Cofilin1; epididymis secretory protein Li 15; nemaline myopathy type 7; p18
Gene Aliases: AA959946; CFL; CFL1; CFL2; Cof; cofilin; HEL-S-15; n-cofilin; NEM7
UniProt ID: (Human) P23528, (Human) Q9Y281, (Rat) P45592, (Mouse) P18760, (Mouse) P45591
Entrez Gene ID: (Human) 1072, (Human) 1073, (Rat) 29271, (Mouse) 12631, (Rat) 366624, (Mouse) 12632
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support