HERC6 Polyclonal Antibody - FAQs
HERC6 Polyclonal Antibody - FAQs
View additional product information for HERC6 Polyclonal Antibody - FAQs (PA542322)
2 product FAQs found
Do you offer a sample size for HERC6 polyclonal antibody?
Unfortunately, we do not offer sample sizes for HER6 polyclonal antibodies (Cat. No. PA5-57426, BS-15463R​, and PA5-57426)​.
However, if you would like to test the antibody to see if it will work for your application, we would recommend enrolling in our Aspire Member Program (www.thermofisher.com/aspire). It is a free program and as an Aspire member, you can receive a free full-size product once every membership year. Fortunately, the HERC6 Polyclonal Antibody (Cat. No. PA5-57426) is part of our trial catalog.
After enrolling in the Aspire Member Program, please allow 24-48 hrs for our Aspire Team to validate your account. Once your account has been validated, you can order your trial product through our Aspire Member website. First log into your account on our webpage (www.thermofisher.com). Then go to Account > Aspire Member Program > Trial products. From here, you can search our catalog of nearly 20,000 full-sized products to try for free.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can I use HERC6 Polyclonal Antibody (Cat. No. PA5-57426) for western blot detection?
Unfortunately, this antibody has not been internally validated for western blot detection, and therefore we cannot guarantee that it will work for this application. We do have two other HERC6 polyclonal antibodies (Cat. No. PA5-42322 and BS-15463R) that have been validated for western blot detection.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.