What was the pH of the buffer used for validating the ASCL2 Polyclonal Antibody (Cat. No. PA5-47852)?
The pH of the buffer used for validating the ASCL2 Polyclonal Antibody (Cat. No. PA5-47852) was pH 9-9.5.
Search
View additional product information for ASCL2 Polyclonal Antibody - FAQs (PA547852)
1 product FAQs found