Have you tested the CD55 Monoclonal Antibody (583905) (Cat. No. MA5-24264) in ICC/IF application?
Unfortunately, this antibody has not been internally verified for ICC/IF, and therefore we cannot guarantee that it will work for this application.
Alternatively, we offer CD55 Recombinant Rabbit Monoclonal Antibody (76) (Cat. No. MA5-29678) and CD55 Polyclonal Antibody (Cat. No. PA5-88462) that target mouse and have been verified for ICC/IF.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Alternatively, we offer CD55 Recombinant Rabbit Monoclonal Antibody (76) (Cat. No. MA5-29678) and CD55 Polyclonal Antibody (Cat. No. PA5-88462) that target mouse and have been verified for ICC/IF.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.