Power Blotter Cassette
Invitrogen™
Power Blotter Cassette
The Invitrogen Power Blotter Cassette is part of the Power Blotter System designed specifically for rapid semi-dry transfer of proteinsRead more
Have Questions?
Catalog NumberQuantity
PB00021 cassette
Catalog number PB0002
Price (USD)
1,353.65
Online exclusive
1,456.00
Save 102.35
Each
Quantity:
1 cassette
Price (USD)
1,353.65
Online exclusive
1,456.00
Save 102.35
Each
The Invitrogen Power Blotter Cassette is part of the Power Blotter System designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter Cassette can simultaneously transfer up to two mini-size or one midi-size gel. This item is made available as a replacement or additional cassette for the Power Blotter Station.
The Power Blotter Cassette can be used with Power Blotter Select Transfer Stacks for rapid transfers, the Power Blotter 1-Step Transfer Buffer for semi-dry transfers, or alternatively with traditional buffers for longer semi-dry transfers.
Learn more about the Power Blotter System ›
Related products
Power Blotter Cassette XL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Membrane CompatibilityNitrocellulose, PVDF
Mode of TransferSemi-Dry
Quantity1 cassette
Shipping ConditionRoom Temperature
Throughput1 Midi or 2 Mini Gels
For Use With (Application)Western Blot
For Use With (Equipment)PB0010 Power Blotter Station
Product LinePower Blotter
TypeBlotter Cassette
Unit SizeEach
Frequently asked questions (FAQs)
What is the difference between the Power Blotter System and the Power Blotter XL System?
The Power Blotter System (Cat. No. PB0012) comes with the Power Blotter Station (Cat. No. PB0010) plus Power Blotter Cassette (Cat. No. PB0002). The Power Blotter System can transfer one midi or up to two mini gels at the same time. The Power Blotter XL System (Cat. No. PB0013) on the other hand comes with the Power Blotter Station (Cat. No. PB0010) plus Power Blotter XL Cassette (Cat. No. PB0003). The Power Blotter XL System can transfer up to two midi or four mini gels at the same time.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Is it possible to do gel staining with the Power Blotter System/Power Blotter XL System?
Even though the Power Blotter System/Power Blotter XL System does not have the staining program, this can be introduced. Please see the user guide for our Pierce Mini/Midi Gel Power Staining Kit (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017799_22839_22840_powerstain_midi_minigel_UG.pdf) for how to create a Custom Staining Method.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What is the material used for the anode and cathode plates of the Power Blotter Cassette/Power Blotter Cassette XL?
The top (cathode) is stainless steel and the bottom (anode) is platinum-coated stainless steel.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
The central European or UK power cord for the Power Blotter System/Power Blotter XL System does not work in Switzerland. Do you offer a Swiss power cord?
Yes, you may purchase the Universal Cable Kit (Cat. No. 4377117).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
I have lost the power cord for the Power Blotter Station. Is it sold as a stand-alone item?
Here are the SKUs for the power cord. Please order the one appropriate for your region:
- Cat. No. 84858: Power Cord with C13 Connector, North America
- Cat. No. 84859: Power Cord with C13 Connector, Continental Europe and South Korea
- Cat. No. 84860: Power Cord with C13 Connector, United Kingdom and Singapore
- Cat. No. 84855: Chinese 10 Amp Cordset w/ C13 Connector
- Cat. No. 84856: Japanese 15 Amp Cordset w/ C13 Connector
- Cat. No. 84857: Australian 10 Amp Cordset w/ C13 Connector
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.