Power Blotter Select Transfer Stacks, PVDF, mini
Invitrogen™

Power Blotter Select Transfer Stacks, PVDF, mini

The Invitrogen Power Blotter Select Transfer Stacks are used to transfer proteins using the Power Blotter System. They are disposable,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSufficient For
PB521010 Blots
PB524040 Blots
Catalog number PB5210
Price (USD)
257.00
Each
Sufficient For:
10 Blots
Price (USD)
257.00
Each
The Invitrogen Power Blotter Select Transfer Stacks are used to transfer proteins using the Power Blotter System. They are disposable, pre-assembled stacks that come with an integrated PVDF transfer membrane for dry blotting of proteins from polyacrylamide gels. Each Power Blotter Select Transfer Stack contains filter paper, membrane, and the appropriate cathode and anode buffers incorporated into a solid gel matrix to allow fast, reliable transfer of proteins.

Features of the Power Blotter Select Transfer Stacks:
• Rapid transfer—the ready-to-use transfer stack takes little setup time and protein transfers take place in as little as seven minutes
• Efficient and even transfer—stacks are made with high-quality membranes that provide high binding efficiency, uniform transfer, and high detection sensitivity
• Increased reproducibility—preassembled stacks provide consistent performance

Learn more about the use of the Power Blotter Select Transfer Stacks in Power Blotter System

PVDF membrane
The PVDF membrane (0.2 μm pore size, low fluorescence) in the stack has higher binding capacity than nitrocellulose. It is pre-activated and ready for use without any pretreatment with alcohols. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. The proteins bind to the membrane through hydrophobic interactions. The protein-binding capacity is 240 μg/cm2.

Mini transfer stacks
The mini-size (8 cm x 8 cm) Power Blotter Select Stacks can be used with all Invitrogen Novex, Bolt, and NuPAGE mini-size gels or equivalent.

Related products
Power Blotter Select Transfer Stacks, PVDF, regular size
Power Blotter Select Transfer Stacks, nitrocellulose, mini size
Power Blotter Select Transfer Stacks, nitrocellulose, regular size

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 stacks
Shipping ConditionRoom Temperature
Dimensions (LxW)8 cm x 8 cm
FormatTransfer Stack
Length (Metric)8 cm
MaterialPVDF
Pore Size0.2 μm
Product LinePower Blotter
Sufficient For10 Blots
Width (Metric)8 cm
Unit SizeEach
Contents & Storage
10 mini-size transfer stacks (8 cm x 8 cm)

Frequently asked questions (FAQs)

What is the difference between the Power Blotter System and the Power Blotter XL System?

The Power Blotter System (Cat. No. PB0012) comes with the Power Blotter Station (Cat. No. PB0010) plus Power Blotter Cassette (Cat. No. PB0002). The Power Blotter System can transfer one midi or up to two mini gels at the same time. The Power Blotter XL System (Cat. No. PB0013) on the other hand comes with the Power Blotter Station (Cat. No. PB0010) plus Power Blotter XL Cassette (Cat. No. PB0003). The Power Blotter XL System can transfer up to two midi or four mini gels at the same time.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Is it possible to do gel staining with the Power Blotter System/Power Blotter XL System?

Even though the Power Blotter System/Power Blotter XL System does not have the staining program, this can be introduced. Please see the user guide for our Pierce Mini/Midi Gel Power Staining Kit (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017799_22839_22840_powerstain_midi_minigel_UG.pdf) for how to create a Custom Staining Method.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the material used for the anode and cathode plates of the Power Blotter Cassette/Power Blotter Cassette XL?

The top (cathode) is stainless steel and the bottom (anode) is platinum-coated stainless steel.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

The central European or UK power cord for the Power Blotter System/Power Blotter XL System does not work in Switzerland. Do you offer a Swiss power cord?

Yes, you may purchase the Universal Cable Kit (Cat. No. 4377117).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

I have lost the power cord for the Power Blotter Station. Is it sold as a stand-alone item?

Here are the SKUs for the power cord. Please order the one appropriate for your region:

- Cat. No. 84858: Power Cord with C13 Connector, North America
- Cat. No. 84859: Power Cord with C13 Connector, Continental Europe and South Korea
- Cat. No. 84860: Power Cord with C13 Connector, United Kingdom and Singapore
- Cat. No. 84855: Chinese 10 Amp Cordset w/ C13 Connector
- Cat. No. 84856: Japanese 15 Amp Cordset w/ C13 Connector
- Cat. No. 84857: Australian 10 Amp Cordset w/ C13 Connector

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.