Normal Goat Serum
Gibco™
Normal Goat Serum
Normal goat serum. Whole serum is obtained from non-immunized (preimmune) healthy goats. The serum is clarified through a 0.8 micronRead more
Have Questions?
Catalog NumberQuantity
PCN50005 mL
Catalog number PCN5000
Price (USD)
95.75
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
95.75
Each
Normal goat serum. Whole serum is obtained from non-immunized (preimmune) healthy goats. The serum is clarified through a 0.8 micron low protein binding filter.
Analyte Specific Reagent. Analytical and performance characteristics are not established.
Specifications
Shipping ConditionDry Ice
SpeciesGoat/Caprine
FormLiquid
Product TypeGoat Serum
Quantity5 mL
SterilitySterile
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).