SURF-CAL™ Particle Size Standards
Thermo Scientific™

SURF-CAL™ Particle Size Standards

Simplify the preparation of calibration wafers with these particle size standards.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Concentration
PD16001.59 μm3 x 108 particles/mL
PD-0470.047 μm3 x 108 particles/mL
PD-047B0.047 μm1 x 10¹° particles/mL
PD-0640.064 μm3 x 108 particles/mL
PD-064B0.064 μm1 x 10¹° particles/mL
PD-0830.083 μm3 x 108 particles/mL
PD-083B0.083 μm1 x 10¹° particles/mL
PD-0920.092 μm3 x 108 particles/mL
PD-092B0.092 μm1 x 10¹° particles/mL
PD-1000.100 μm3 x 108 particles/mL
PD-100B0.100 μm1 x 10¹° particles/mL
PD11001.112 μm3 x 108 particles/mL
PD1100B1.112 μm1 x 10¹° particles/mL
PD-1250.126 μm3 x 108 particles/mL
PD-125B0.126 μm1 x 10¹° particles/mL
PD-1550.155 μm3 x 108 particles/mL
PD-155B0.155 μm1 x 10¹° particles/mL
PD-2000.202 μm3 x 108 particles/mL
PD20002.01 μm3 x 108 particles/mL
PD-200B0.202 μm1 x 10¹° particles/mL
PD-2040.204 μm3 x 108 particles/mL
PD-204B0.204 μm1 x 10¹° particles/mL
PD-2150.220 μm3 x 108 particles/mL
PD-215B0.220 μm1 x 10¹° particles/mL
PD30003.04 μm3 x 108 particles/mL
PD-3050.304 μm3 x 108 particles/mL
PD-305B0.304 μm1 x 10¹° particles/mL
PD-3650.360 μm3 x 108 particles/mL
PD-365B0.360 μm1 x 10¹° particles/mL
PD-5000.498 μm3 x 108 particles/mL
PD-500B0.498 μm1 x 10¹° particles/mL
PD-8000.809 μm3 x 108 particles/mL
PD-800B0.809 μm1 x 10¹° particles/mL
PD-8020.802 μm3 x 108 particles/mL
PD-802B0.802 μm1 x 10¹° particles/mL
Catalog number PD1600
Price (USD)
-
Diameter (Metric):
1.59 μm
Concentration:
3 x 108 particles/mL
Simplify the preparation of calibration wafers with Thermo Scientific™ SURF-CAL™ Particle Size Standards, for calibration and maintenance of Scanning Surface Inspection Systems (SSIS). Available particle sizes correspond to the calibration point sizes required by instrument manufacturers.
Thermo Fisher Scientific™, along with the Semiconductor Industry and SEMI, have established specific particle sizes to be used when calibrating Scanning Surface Inspection Systems. Working with instrument manufacturers, we created the SURF-CAL™ product line to meet SEMI standard guidelines.
  • Available sizes are considered to be critical sizing nodes as defined by the International Technology Roadmap for Semiconductors (ITRS).
  • By depositing SURF-CAL™ NIST™ traceable polystyrene latex spheres (PSL) on specially selected wafers, you can perform periodic calibration checks and compare your scanner with scanners at other locations.
  • You can also assess the performance of your SSIS at critical stages in the manufacturing process.
  • All products are suspended in deionized, filtered water in 50mL bottles at a concentration of 3 × 108 particles per mL.

Note:

Products PD1100 and smaller are also available at 1010 particles per mL for applications using the aid of a Differential Mobility Analyzer (DMA) or other size exclusionary techniques.

Specifications
AdditiveNone
Concentration3 x 108 particles/mL
ContentsPolystyrene microspheres in deionized, filtered water
DescriptionParticles for calibration and maintenance of Scanning Surface Inspection Systems (SSIS)
IncludesCertificate of Calibration and Traceability to NIST
Particle Density1.05g/cm³
Quantity50 mL
Refractive Index1.59 at 589nm (25°C)
Size Distribution0.016µm
StabilityProduct stable for a minimum of 12 months
ColorWhite or Colorless
Diameter (Metric)1.59 μm
For Use With (Application)SSIS Calibration and Maintenance
MaterialPolystyrene
Product TypeSize Standard Particles
Unit SizeEach
Contents & Storage
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.