Product Image
Applied Biosystems™

Pathatrix™ Foam Filters

Pathatrix™ Foam Filters are specially designed single-use filters that fit easily onto the Pathatrix™ Pooling Kit straws and serve to remove particulate matter from pre-enriched food and environmental samples, improving the quality of downstream analysis.
Have Questions?
Catalog NumberQuantity
PFF1000 single-use filters
Catalog number PFF
Price (USD)
369.00
Each
Quantity:
1000 single-use filters
Price (USD)
369.00
Each

Pathatrix™ Foam Filters are specially designed single-use filters that fit easily onto the Pathatrix™ Pooling Kit straws and serve to remove particulate matter from pre-enriched food and environmental samples, improving the quality of downstream analysis.

These filters are sealed in sterile packs of 5, and are designed for use with the Pathatrix 5 Pooling Kit or the Pathatrix™ 10 Pooling Kit.

Specifications
For Use With (Application)Pathatrix™ Pooling Kits (POOL510MLN, POOL1010MLN)
For Use With (Equipment)Pathatrix™ Auto System
Product LinePathatrix
Product TypeFoam Filter
Quantity1000 single-use filters
Shipping ConditionRoom Temperature
SterilitySterile
Unit SizeEach
Contents & Storage
1000 single-use filters in sterile and sealed packs of 5 filters.

Store at room temperature.