IL6 Recombinant Guinea Pig

Catalog number: PGC0064

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant guinea pig IL6 is a bioactive protein. Interleukin 6 (IL6) is a pleotropic cytokine involved in several cellular processes including inflammation, hematopoeisis, and immune response.

ED50 = ≤ 20 pg⁄mL, determined by the dose dependent proliferation of mouse B9 cells.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Bone Research, Immunology, Inflammation, Stem Cell Research
Biological Activity: ED50 = ≤ 20 pg⁄mL
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: IL-6
Mass: 10 µg
Molecular Weight: 20 kDa
Product Line: Gibco®
Product Size: 10 µg
Protein Family: Cytokines & Receptors
Protein Form: Full Length, Ligand, Recombinant
Protein Subtype: Interleukins
Purification: Sequential Chromatography
Species: Guinea Pig
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Store this lyophilized preparation at 2-8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤–20°C. Avoid repeated freeze⁄thaw cycles.

Documents

Manuals & protocols

Recommended products