Gibco™

IL9 Recombinant Human Protein

Catalog number:  PHC0091

Related applications:

Mammalian Cell Culture

Back to top

Description

Recombinant Human Interleukin 9 (IL9) is a bioactive protein intended for use in cell culture applications. IL9 is produced by CD4+ helper cells, and has been shown to be a pleiotropic cytokine impacting lymphoid and mast cell lineages. This is the 40 microgram size.

• High purity—no interference from other proteins or contaminants
• High biological activity—more results with less protein
• Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High purity recombinant bioactive protein
Recombinant Human Interleukin 9 (IL9) is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to get a biologically active protein.

High biological activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays. Recombinant Human Interleukin 9 (IL9) specific activity is determined by the dose-dependent stimulation of the proliferation of human MO7e cells.

Proven compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been a consistent provider of high quality media, reagents, and sera for reliable cell culture.

Related links
See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Activity: 0.2 ng/ml
Bioassay: Proliferation of human MO7e cells
Endotoxin Level: < 1 EU/ml
Expression: Mammalian
Form: Lyophilized
Gene Aliases: IL-9
Mass: 100 µg
Molecular Weight: 48 kDa
Product Size: 100 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand,Recombinant
Protein Subtype: Interleukins
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Research Category: Apoptosis,Oncology,Signal Transduction,Stem Cell Research
Shipping Condition: Dry Ice
Species: Human
Validated Application: Cell Culture

Contents & storage

Store in freezer (-5 to -30°C).