Gibco™

RANTES Recombinant Human Protein

Catalog number:  PHC1051

Related applications:

Mammalian Cell Culture

Back to top

Description

Recombinant RANTES is a bioactive protein intended for use in cell culture applications. RANTES functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. RANTES is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 0.1 ng/µg
Mass: 100 µg
Product Line: Gibco®
Purity or Quality Grade: Carrier-Free
Shipping Condition: Wet Ice
Expression System: E. coli
Form: Lyophilized
Protein Family: Chemokines & Receptors
Protein Form: Recombinant, Ligand
Protein Subtype: Rantes
Research Category: Immunology, Stem Cell Research, Oncology
Species: Human
Validated Application: Cell Culture
Gene ID (Entrez): 6352
Molecular Weight: 7.8 kDa
Purification Method: Sequential Chromatography
Purity: 95 %
Quantity: 100μg
Gene Alias: RANTES

Contents & storage

Store in refrigerator (2–8°C).