SDF1A Recombinant Human Protein

Catalog number: PHC1346

Gibco™  Related applications: Mammalian Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant SDF1A is a bioactive protein intended for use in cell culture applications. SDF1A is a small cytokine that belong to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL-1.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Cardiovascular Research, Immunology, Neurobiology, Stem Cell Research
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: SDF-1α
Gene ID: 6387
Mass: 25 µg
Molecular Weight: 8 kDa
Product Line: Gibco®
Product Size: 25 µg
Protein Family: Chemokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: SDFs (Stromal Cell-Derived Factors)
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Human
Validated Application: Cell Culture
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products