GM-CSF Recombinant Human - FAQs
GM-CSF Recombinant Human - FAQs
View additional product information for GM-CSF Recombinant Human - FAQs (PHC2014)
1 product FAQs found
How was the protein concentration of the recombinant swine GM-CSF determined?
We determined the protein concentration of our recombinant swine GM-CSF by measuring the OD280. Based on the amino acid composition of the protein, the conversion factor between OD and protein concentration is as follows: 1 OD = 0.9 mg/mL protein.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.