Specifications
ClassificationCarrier-Free
Endotoxin Level< 0.1 ng/μg
Expression SystemMammalian
Gene AliasIL-29
Mass100 μg
Molecular Weight (g/mol)35 kDa
Product LineGibco
Protein FamilyCytokines & Receptors
Protein FormRecombinant, Ligand
Protein SubtypeInterleukins
Purification MethodSequential Chromatography
Purity or Quality Grade>95% by SDS-PAGE, >95% by SDS-PAGE
Quantity100 μg
ReactivityHuman
Research CategoryVirology, Immunology
Shipping ConditionWet Ice
For Use With (Application)Cell Culture
FormLyophilized
RecombinantRecombinant
SpeciesHuman
Unit Size100 µg