IGFBP6 Recombinant Human Protein

Catalog number: PHC9624

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Human Insulin-like Growth Factor Binding Protein 6 (IGFBP6) is a bioactive protein intended for use in cell culture applications. IGFBP6 binds IGF-1 and II, prolonging these growth factors’ half life and altering the way these growth factors interact with cells surface receptors. This is the 10 microgram size.

• High purity—Minimizes interference from other proteins or contaminants
• High biological activity—More results with less protein
• Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High Purity Recombinant Bioactive Protein
Recombinant Human Insulin-like Growth Factor Binding Protein 6 (IGFBP6) is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein.

High Biological Activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays. Recombinant Human Insulin-like Growth Factor Binding Protein 6 (IGFBP6) specific activity is determined the dose dependent inhibition of IGF-II induced proliferation of MCF-7 cells.

Proven Compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been a consistent provider of high quality media, reagents, and sera for reliable cell culture.

Related Links
See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Apoptosis, Oncology
Activity: 0.4 µg/ml
Bioassay: inhibition of IGF-II induced proliferation of MCF-7 cells
Endotoxin Level: < 1 EU/ml
Expression: Mammalian
Form: Lyophilized
Gene Aliases: IGFBP-6
Mass: 10 µg
Molecular Weight: 22.6 kDa
Product Line: Gibco®
Product Size: 10 µg
Protein Family: Growth Factors & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: IGFs (Insulin-Like Growth Factors)
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Store at 2 to 8°C

Documents

Manuals & protocols

Recommended products