Get out of the cold & warm up to new Gibco BenchStable Media! Optimized for storage at room temperature »
Gibco™

TGFB2 Recombinant Human Protein

Catalog number:  PHG9124

Related applications:

Cell Analysis

| Cell Culture

| Stem Cell Research

| Stem Cell Research

Back to top

Description

Recombinant Human Transforming Growth Factor β 2 (TGFB2) is a bioactive protein intended for use in cell culture applications. TGFB2 binds the transforming growth factor-β receptor and plays a role in regulation of cell growth and proliferation. It may be involved in mesenchymal-epithelial cell interactions during development.

High purity—no interference from other proteins or contaminants
High biological activity—more results with less protein
Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High Purity Recombinant Bioactive Protein
Recombinant Human Transforming Growth Factor β 2 is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.

High Biological Activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Transforming Growth Factor β 2 (TGFB2) specific activity is determined by its ability to inhibit the mouse IL-4 dependent proliferation of mouse HT-2 cells.

Proven Compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.

See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Activity: 1 ng⁄ml
Bioassay: Inhibition of Proliferation of HT2 cells
Endotoxin Level: < 1 EU/ml
Product Line: Gibco®
Purity or Quality Grade: Carrier-Free
Shipping Condition: Wet Ice
Expression System: Mammalian
Form: Lyophilized
Protein Family: Growth Factors & Receptors
Protein Form: Recombinant, Ligand
Protein Subtype: TGFs (Transforming Growth Factors)
Research Category: Immunology, Neurobiology, Oncology, Stem Cell Research, Cardiovascular Research, Differentiation, Bone Research
Species: Human
Validated Application: Cell Culture
Molecular Weight: 25 kDa
Purification Method: Sequential Chromatography
Purity: 95 %
Quantity: 10μg
Gene Alias: TGF-β2
Protein Length: 112 aa

Contents & storage

Store at 2-8° C