Product Image
Invitrogen™

WORTMANNIN

WORTMANNIN
Have Questions?
Catalog NumberQuantity
PHZ13011 mg
Catalog number PHZ1301
Price (USD)
191.00
Each
Quantity:
1 mg
Price (USD)
191.00
Each

Wortmannin, a fungal metabolite, is a potent and selective inhibitor of phosphatidylinositol 3-kinase.

Format/Formulation: Lyophilized

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Gene ID (Entrez)5290, 5291, 5293, 5294
Inhibitor PropertiesChemical
InhibitorsKinase Inhibitor (PI 3-Kinase Inhibitor), Kinase Inhibitor
Molecular Weight (g/mol)428.4
Product TypeWortmannin
Purification MethodTLC
Quantity1 mg
Shipping ConditionDry Ice
Specific Inhibit or TargetPI3-Kinase
SterilityNon-sterile
FormSolid
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).