PINspector+ Pin Hole Detector
PINspector+ Pin Hole Detector
PINspector+ Pin Hole Detector
Thermo Scientific™
PINspector+ Pin Hole Detector
The Thermo Scientific PINspector+ provides contactless monitoring of flat sheet for defects that could compromise the integrity of the barrier between perishable goods and the outside environment.
Have Questions?
Número de catálogoDescripción
PINSPECTORPINspector+ Detector de agujeros muy pequeños
Número de catálogo PINSPECTOR
Precio (MXN)
-
Descripción:
PINspector+ Detector de agujeros muy pequeños
Pin holes and edge cracks compromise overall sheet quality resulting in downgraded material or scrap.The Thermo Scientific™ PINspector+ provides contactless monitoring of flat sheet for such defects. The instrument can detect holes as small as 5 microns on sheet speeds up to 2000 meters per minute. This is critical for food canning applications where the smallest hole can spoil the contents and put the end consumer's health at risk. The PINspector+ can also be configured to show width measurement and provide additional information on strip quality.
The Thermo Scientific PINspector+ Pin Hole Detector is recommended for coating lines, slitting lines and other process lines where sheet integrity is critical.
  • Hole detection as small as 5 microns
  • High speed detection for line-speeds as fast as 2000 meters per minute
  • Excellent hole position information: 1 mm in rolling direction, 6 mm in cross strip direction
  • Configurable hole size discriminator
  • No edge masks – completely contactless monitoring
  • C-frame configuration with optional manual or motorized roll off mechanics
  • Edge crack detection for cracks as small as 200 microns 
  • TCP/IP connectivity
  • Optional printed coil reports
  • Optional width measurement  
Especificaciones
DescripciónPINspector+ Detector de agujeros muy pequeños
AplicacionesPin Hole Detection
Unit SizeEach