RANTES Recombinant Mouse Protein

Catalog number: PMC1056

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant RANTES is a bioactive protein intended for use in cell culture applications. RANTES functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. RANTES is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Oncology, Stem Cell Research
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: RANTES
Gene ID: 20304
Mass: 25 µg
Molecular Weight: 8 kDa
Product Line: Gibco®
Product Size: 25 µg
Protein Family: Chemokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: Rantes
Purification: Sequential Chromatography
Purity: 95 %
Species: Mouse
Validated Application: Cell Culture
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products