Get out of the cold & warm up to new Gibco BenchStable Media! Optimized for storage at room temperature »
Gibco™

TPO Recombinant Mouse Protein

Catalog number:  PMC1144

Related applications:

Mammalian Cell Culture

| Stem Cell Culture

Back to top

Description

Recombinant TPO is a bioactive protein intended for use in cell culture applications. TPO is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. This protein is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 0.1 ng/µg
Mass: 10 µg
Product Line: Gibco®
Purity or Quality Grade: Carrier-Free
Shipping Condition: Approved for shipment on Wet or Dry Ice
Expression System: E. coli
Form: Lyophilized
Protein Family: Growth Factors & Receptors
Protein Form: Recombinant, Ligand
Protein Subtype: TPO
Research Category: Endocrinology, Stem Cell Research, Angiogenesis
Species: Mouse
Validated Application: Cell Culture
Gene ID (Entrez): 22018
Molecular Weight: 17 kDa
Purification Method: Sequential Chromatography
Purity: 95 %
Quantity: 10μg
Gene Alias: TPO

Contents & storage

Store in refrigerator (2–8°C).