Get out of the cold & warm up to new Gibco BenchStable Media! Optimized for storage at room temperature »
Gibco™

TPO Recombinant Mouse Protein

Catalog number:  PMC1145

Related applications:

Mammalian Cell Culture

| Stem Cell Culture

Back to top

Description

Recombinant TPO is a bioactive protein intended for use in cell culture applications. TPO is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. This protein is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: TPO
Gene ID: 22018
Mass: 25 µg
Molecular Weight: 17 kDa
Product Line: Gibco®
Product Size: 25 µg
Protein Family: Growth Factors & Receptors
Protein Form: Ligand,Recombinant
Protein Subtype: TPO
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Research Category: Angiogenesis,Endocrinology,Stem Cell Research
Shipping Condition: Approved for shipment on Wet or Dry Ice
Species: Mouse
Validated Application: Cell Culture

Contents & storage

Store in refrigerator (2–8°C).