Gibco™

G-CSF Recombinant Mouse Protein

Back to top

Description

Recombinant G-CSF is a bioactive protein intended for use in cell culture applications. G-CSF stimulates the proliferation and differentiation of hematopoietic progenitor cells.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 0.1 ng/µg
Mass: 25 µg
Product Line: Gibco®
Purity or Quality Grade: Carrier-Free
Shipping Condition: Approved for shipment on Wet or Dry Ice
Expression System: E. coli
Form: Lyophilized
Protein Family: Cytokines & Receptors
Protein Form: Recombinant, Ligand
Protein Subtype: GM-CSF, G-CSF, & M-CSF
Research Category: Signal Transduction, Oncology, Stem Cell Research, Differentiation, Drug Development
Species: Mouse
Validated Application: Cell Culture
Gene ID (Entrez): 12985
Molecular Weight: 19 kDa
Purification Method: Sequential Chromatography
Purity: 95 %
Quantity: 25μg
Gene Alias: G-CSF

Contents & storage

Store in refrigerator (2–8°C).