Pre-Preserved Environmental Sample Containers
Pre-Preserved Environmental Sample Containers
Pre-Preserved Environmental Sample Containers
Pre-Preserved Environmental Sample Containers
Pre-Preserved Environmental Sample Containers
Thermo Scientific™

Pre-Preserved Environmental Sample Containers

Save the time, tedium and chemical contact associated with diluting and aliquoting chemical preservatives by using our chemically-preserved sample containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorPreservative
PP140-40CEP.2HAClear0.2 mL 1:1 Hydrochloric Acid
PP140-40CEP.5HAClear0.5 mL 1:1 Hydrochloric Acid
PP141-40AEP.2HAAmber0.2 mL 1:1 Hydrochloric Acid
PP141-40AEP.5HAAmber0.5 mL 1:1 Hydrochloric Acid
Catalog number PP140-40CEP.2HA
Price (USD)
-
Color:
Clear
Preservative:
0.2 mL 1:1 Hydrochloric Acid

Simplify field preservation with Thermo Scientific™ Chemically-Preserved Environmental Sample Containers and enjoy peace of mind as well as convenience. Using certified containers and chemicals reduces the risk of container and preservative contamination. Let us save you the time, tedium and chemical contact associated with diluting and aliquoting chemical preservatives to prepare environmental sample container sets. Choose from our standard offering of chemically-preserved containers, or let us custom-create container/preservative combos according to your personal analytical needs.

  • Standard Thermo Scientific Certified sample containers chemically-preserved with certified chemical preservatives for environmental sample analyses
  • Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification for highway and rail transport

Ordering Information:

Custom preserved containers available by special order:

  • Choose a standard certified Thermo Scientific environmental sample container
  • Choose a chemical from our standard chemical list
  • Specify the volume and concentration of chemical to aliquot into each container

Products available only in the US

Includes:

Certified sample containers dosed with chemical preservative; certificate of analysis for container and preservative, individually barcoded containers traceable to certs, sample analysis labels unattached for optional use, hazardous material compliant packaging. MSDS available upon request.

Specifications
Certifications/ComplianceCertified to meet the EPA Performance Based Specifications for volatile organic compounds
Closure Size24-414
Diameter (English) Outer1.1 in.
Height (English)3.75 in.
Height (Metric)95 mm
LevelCertified
No. per Case72
Preservative0.2 mL 1:1 Hydrochloric Acid
Product TypeEnvironmental Sample Container
Thickness (Metric)3.17 mm
Volume (English)1 oz.
Volume (Metric) Total40 mL
Closure TypeSepta Cap
ColorClear
Diameter (Metric) Outer28 mm
MaterialGlass (Type 1)
Volume (Metric)40 mL
Unit SizeEach