Aluminum Support, tube/plate rack, 96-well
Thermo Scientific™
Aluminum Support, tube/plate rack, 96-well
The Thermo Scientific 96-well Aluminium Support Rack provides optimal support for heat sealing and automated dispensing for all 0.2mL PCRRead more
Have Questions?
Catalog NumberQuantity
PRK19631 rack
Catalog number PRK1963
Price (USD)
403.00
Each
Quantity:
1 rack
Price (USD)
403.00
Each
The Thermo Scientific 96-well Aluminium Support Rack provides optimal support for heat sealing and automated dispensing for all 0.2mL PCR tubes and standard 96-well PCR plates, as well as 24-well Piko PCR Plates and Plate/Frame assemblies.
Features
  • V-shaped wells give optimal support for ultra-thin walls for applying and removing sealers
  • Made of blue aluminum
  • Industry standard footprint fits in robotic workstations, enabling automated pipetting with Piko PCR plates
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Quantity1 rack
ColorBlue
HoldsMicroplates (-well)
MaterialAluminum
Product TypeMicroplate Rack
ShapeRectangular
Unit SizeEach