PowerEase™ Touch 350W Power Supply, 230 VAC, 1 each - FAQs
PowerEase™ Touch 350W Power Supply, 230 VAC, 1 each - FAQs
View additional product information for PowerEase™ Touch Power Supply - FAQs (PS0351)
4 product FAQs found
Is the PowerEase Touch Power Supply (Cat. No. PS0351) compatible with the Owl gel Systems?
Yes, it is compatible with the Owl Gel Systems. The following ones can be used with the PowerEase power supplies with no need for adaptors:
    Owl EasyCast B1 Mini Gel Electrophor. Sys. (Cat. No. B1-BP)
    Owl A2 Large Gel Sys. (Cat. No. A2)
    Owl EasyCast B3 Gel Sys. with Built-In Recirculat. (Cat. No. B3).
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
Can the four sets of output terminals on the PowerEase Touch 350W Power Supply be used with four different programs?
The four sets of output terminals allow simultaneous running of multiple electrophoresis units, but they will run under the same program.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Which applications can be run using the PowerEase Touch 350 W Power Supply?
The PowerEase Touch 350W Power Supply is ideal for high current applications like high throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I use the PowerEase Touch 120W Power Supply for transferring gels with the SureLock Tandem Midi Blot Module, Mini Gel Blot Module, or Novex Semi-Dry Blotter?
No, most transfer applications require high current and therefore, we recommend using a high current power supply like PowerEase Touch 350W Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.