PowerEase™ Touch 350W Power Supply, 230 VAC, China, 1 each - FAQs
PowerEase™ Touch 350W Power Supply, 230 VAC, China, 1 each - FAQs
View additional product information for PowerEase™ Touch Power Supply - FAQs (PS0353)
6 product FAQs found
Which applications can be run using the PowerEase Touch 350 W Power Supply?
The PowerEase Touch 350W Power Supply is ideal for high current applications like high throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I use the PowerEase Touch 120W Power Supply for transferring gels with the SureLock Tandem Midi Blot Module, Mini Gel Blot Module, or Novex Semi-Dry Blotter?
No, most transfer applications require high current and therefore, we recommend using a high current power supply like PowerEase Touch 350W Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I use the PowerEase Touch 120W or PowerEase Touch 350W Power Supply to run isoelectric focusing (IEF) gels?
The voltage required for running of IEF gels cannot be reached with the PowerEase Touch 120W or the PowerEase Touch 350W Power Supply. We recommend using the PowerEase Touch 600W Power Supply or PowerEase Touch HV Power Supply for running IEF gels.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How many gels can I simultaneously transfer using the PowerEase Touch 350W Power Supply?
The PowerEase Touch 350W Power Supply can be used for simultaneous transfer of 8 mini gels or 4 midi gels.
How many gels can I simultaneously run using the PowerEase Touch 350W Power Supply?
The PowerEase Touch 350W Power Supply can be used for simultaneous running of 12 mini gels or 8 midi gels.