Get out of the cold & warm up to new Gibco BenchStable Media! Optimized for storage at room temperature »
Gibco™

IL4 Recombinant Swine Protein

Catalog number:  PSC0044

Related applications:

Mammalian Cell Culture

Back to top

Description

Recombinant IL4 is a bioactive protein intended for use in cell culture applications. IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: IL-4
Gene ID: 397225
Mass: 10 µg
Molecular Weight: 12.5 kDa
Product Line: Gibco®
Product Size: 10 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand,Recombinant
Protein Subtype: Interleukins
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Research Category: Immunology,Inflammation,Stem Cell Research
Shipping Condition: Approved for shipment on Wet or Dry Ice
Species: Swine⁄Porcine
Validated Application: Cell Culture

Contents & storage

Store in refrigerator (2–8°C).