PowerEase™ Touch 120W Power Supply and Mini Gel Tank Welcome Pack
Invitrogen™

PowerEase™ Touch 120W Power Supply and Mini Gel Tank Welcome Pack

The PowerEase Touch 120W Power Supply* and Mini Gel Tank Welcome Pack contains:• PowerEase Touch 120W Power Supply*• PowerEase TouchRead more
Have Questions?
Catalog NumberQuantity
PSC120M1 pack
Catalog number PSC120M
Price (USD)
946.40
Special offer
Online exclusive
Ends: 04-Dec-2026
1,456.00
Save 509.60
Each
Quantity:
1 pack
Price (USD)
946.40
Special offer
Online exclusive
Ends: 04-Dec-2026
1,456.00
Save 509.60
Each
The PowerEase Touch 120W Power Supply* and Mini Gel Tank Welcome Pack contains:

• PowerEase Touch 120W Power Supply*
• PowerEase Touch 120W Power Supply (230 VAC)*
• PowerEase Touch 120W Power Supply (230 VAC) (Australia)*
• PowerEase Touch 120W Power Supply (230 VAC) (China)*
• Mini Gel Tank

*dependent on region

About the PowerEase Touch 120W Power Supply
The Invitrogen PowerEase Touch 120W Power Supply is a medium-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for DNA/RNA electrophoresis, SDS-PAGE, and native PAGE. The PowerEase Touch 120W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the Mini Gel Tank
The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.
Specifications
Ambient Temperature (English)Room Temperature
Dimensions (L x W x H)30.3 inch x 23 inch x 10.2 inch
Display4.3 inch backlit LCD
Max. Current500 mA
No. of Jacks4 (4 positive voltage and 4 negative voltage)
Quantity1 pack
Shipping ConditionRoom temperature
StackableYes
TimerUp to 999 min./step
WarrantyOne year
Wattage120 watts
Weight2.1 kg
For Use With (Application)DNA/RNA electrophoresis, SDS-PAGE, and native PAGE
Gel SizeMini
Max. Voltage300 VDC
Product LinePowerEase
Unit SizeEach