GM-CSF Recombinant Swine Protein

Catalog number: PSC2014

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications. Recombinant swine GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Drug Development, Oncology, Signal Transduction, Stem Cell Research
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: GM-CSF
Gene ID: 397208
Mass: 10 µg
Molecular Weight: 14 kDa
Product Line: Gibco®
Product Size: 10 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: GM-CSF, G-CSF, & M-CSF
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Swine⁄Porcine
Validated Application: Cell Culture
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products