PowerEase™ Touch 350W Power Supply and and Mini Gel Tank with Mini Blot Module Set Welcome Pack - FAQs
PowerEase™ Touch 350W Power Supply and and Mini Gel Tank with Mini Blot Module Set Welcome Pack - FAQs
View additional product information for PowerEase™ Touch 350W Power Supply and and Mini Gel Tank with Mini Blot Module Set Welcome Pack - FAQs (PSC350MB)
3 product FAQs found
Can I use the PowerEase Touch 120W or PowerEase Touch 350W Power Supply to run isoelectric focusing (IEF) gels?
The voltage required for running of IEF gels cannot be reached with the PowerEase Touch 120W or the PowerEase Touch 350W Power Supply. We recommend using the PowerEase Touch 600W Power Supply or PowerEase Touch HV Power Supply for running IEF gels.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How many gels can I simultaneously transfer using the PowerEase Touch 350W Power Supply?
The PowerEase Touch 350W Power Supply can be used for simultaneous transfer of 8 mini gels or 4 midi gels.
How many gels can I simultaneously run using the PowerEase Touch 350W Power Supply?
The PowerEase Touch 350W Power Supply can be used for simultaneous running of 12 mini gels or 8 midi gels.