PowerEase™ Touch 350W Power Supply and SureLock™ Tandem Midi Gel Tank Welcome Pack
Invitrogen™

PowerEase™ Touch 350W Power Supply and SureLock™ Tandem Midi Gel Tank Welcome Pack

The PowerEase Touch 350W Power Supply* and SureLock Tandem Midi Gel Tank Welcome Pack contains: • PowerEase Touch 350W PowerRead more
Have Questions?
Catalog NumberQuantity
PSC350TT1 pack
Catalog number PSC350TT
Price (USD)
1,459.25
Special offer
Online exclusive
Ends: 04-Dec-2026
2,245.00
Save 785.75
Each
Quantity:
1 pack
Price (USD)
1,459.25
Special offer
Online exclusive
Ends: 04-Dec-2026
2,245.00
Save 785.75
Each
The PowerEase Touch 350W Power Supply* and SureLock Tandem Midi Gel Tank Welcome Pack contains:

• PowerEase Touch 350W Power Supply (115 VAC)* or PowerEase Touch 350W Power Supply (230 VAC)* or PowerEase Touch 350W Power Supply (230 VAC) (Australia)* or PowerEase Touch 350W Power Supply (230 VAC) (China)*
• SureLock Tandem Midi Gel Tank

*dependent on region

About the PowerEase Touch 350W Power Supply
The Invitrogen PowerEase Touch 350W Power Supply is a high-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for high current applications like high throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting. The PowerEase Touch 350W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the SureLock Tandem Midi Gel Tank
The Invitrogen SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a setup time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.

Specifications
Ambient Temperature (English)Room Temperature
Dimensions (L x W x H)30.3 inch x 23 inch x 10.2 inch
Display4.3 inch backlit LCD
Max. Current3 A
No. of Jacks4 (4 positive voltage and 4 negative voltage)
Quantity1 pack
Shipping ConditionRoom Temperature
StackableYes
TimerUp to 999 min./step
WarrantyOne year
Wattage350 watts
Weight2.1 kg
For Use With (Application)High throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE and western blotting
Gel SizeMidi
Max. Voltage300 VDC
Product LinePowerEase
Unit SizeEach