5X Kinase Buffer A
Actual product may vary
Invitrogen™

5X Kinase Buffer A

The 5X Kinase Buffer A is a concentrated general kinase assay buffer that can be used with a variety ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV31894 mL
PV6135100 ml
Catalog number PV3189
Price (USD)
230.65
Online exclusive
251.00
Save 20.35
Each
Quantity:
4 mL
Price (USD)
230.65
Online exclusive
251.00
Save 20.35
Each
The 5X Kinase Buffer A is a concentrated general kinase assay buffer that can be used with a variety of assay technologies such as, LanthaScreen™, Adapta™, and Z’-LYTE™ assays.

Ready to use and validated with LanthaScreen™, Adapta™, and Z’LYTE™ assays

• Optimized 5X concentrated formulation
• Simplified handling with room temperature storage

Easily Screen for Kinase Targets using an Invitrogen™ Platform
For more information on the kinase assays that can be used with our 5X Kinase Buffer A see the following links:

LanthaScreen™ Eu Binding Assay
LanthaScreen™ Activity Assay
Adapta™ Universal Kinase Assay
Z´-LYTE™ Kinase Assay Kits

Save Freezer Space
The 5X Kinase Buffer can be stored at room temperature, saving you time and precious freezer space.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialKinase Buffer A
Includes5X Kinase Buffer A may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).
Concentration5X
For Use With (Application)Pharma & Biopharma; Protein Assays and Analysis
Physical FormLiquid
Quantity4 mL
Unit SizeEach