Human ROCK2, GST Tag Recombinant Protein - FAQs
Human ROCK2, GST Tag Recombinant Protein - FAQs
View additional product information for Human ROCK2, GST Tag Recombinant Protein - FAQs (PV4048, PV3759, PR7116B)
1 product FAQs found
What is the amino acid sequence for the ROCK2 Recombinant Human Protein?
The amino acid sequence for ROCK2 Recombinant Human Protein can be found in the Certificate of Analysis for your specific lot number, see example here for Lot number 2112471C:
https://assets.thermofisher.com/TFS-Assets/LSG/certificate/Certificates-of-Analysis/COA_PV4048_2112471C.pdf