Product Image
Thermo Scientific™
Tampón corregulador TR-FRET C
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Have Questions?
Número de catálogoCantidad
PV441925 mL
Número de catálogo PV4419
Precio (MXN)
-
Cantidad:
25 mL
Un tampón receptor nuclear especializado que se puede utilizar con la colección de Invitrogen de proteínas y ensayos de receptores nucleares purificados.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (equipo)Ensayos de interacción del cofactor del receptor nuclear LanthaScreen™
Cantidad25 mL
Condiciones de envíoAprobado para su envío a temperatura ambiente o en hielo húmedo o seco
Unit SizeEach
Contenido y almacenamiento
El tampón C corregulador TR-FRET puede enviarse congelado o a temperatura ambiente. Conservar a 4°C.
Preguntas frecuentes
I have no assay window in my TR-FRET assay. What is the reason?
The single most common reason that a TR-FRET assay fails is that an incorrect choice of emission filters was made. Unlike other fluorescent assays, the filters used in a TR-FRET assay must be exactly those that are recommended for your instrument as the emission filter choice can make or break the assay. The excitation filter has more of an impact on the assay window. Please refer to our instrument setup guides in our instrument compatibility portal (https://www.thermofisher.com/instrumentsetup). If your instrument is not listed there, please contact Drug Discovery Technical Support at drugdiscoverytech@thermofisher.com.
Note: Test your microplate reader's TR-FRET setup before you begin any work with your assay. Using reagents that you have already purchased for your assay, you can test the microplate reader's TR-FRET setup. Please refer to the Terbium (Tb) Assay (http://tools.thermofisher.com/content/sfs/manuals/LanthaScreen_Tb_Instrument_Control_Application_Note17Sep10.pdf) and Europium (Eu) Assay (http://tools.thermofisher.com/content/sfs/manuals/Eu_Instrument_Control_AppNote_12Nov10.pdf) Application Notes for plate reader setup.
Find additional tips, troubleshooting help, and resources within our Drug Discovery & Development Support Center.