In an Adapta assay, how do I calculate IC₅₀s from the Emission Ratio?
The response of Adapta assays to ADP formation is not linear. Therefore, the concentration of inhibitor required to cause a 50% decrease in the assay signal, measured as the emission ratio of the 665 nm to 615 nm RFU is NOT equivalent to the IC₅₀ value of the inhibitor. The EC₅₀ will be right-shifted compared to the IC₅₀.
As more inhibitor is present, less ADP is formed and more ADP Tracer stays bound to the antibody. Therefore, as the concentration of inhibitor increase, the signal increases.
In an assay in which the signal response is linear (or nearly linear) with respect to product formation, such as the LanthaScreen Eu Kinase Binding Assays, Z-LYTE Assays and others, the concentration of inhibitor that results in a 50% decrease in the assay signal, the EC₅₀, as determined by the emission ratio is equivalent to the IC₅₀ value for that inhibitor.
Since the response of the Adapta Universal Kinase Assay to ADP formation is not linear, the concentration of inhibitor required to cause a 50% decrease in the Emission Ratio, the EC₅₀, is NOT equivalent to the IC₅₀ value for the inhibitor.
To convert an inhibitor EC₅₀ value to an IC₅₀ value, use an ATP-ADP standard curve to correct for the non-linear response of the Adapta assay. ADP and ATP are provided in the kit. The curve is generated at constant total ATP plus ADP with known amounts of ATP and ADP.
Note: The substrate should be included in this titration.
Please refer to the Adapta Universal Kinase Assay User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/adapta_userguide_man.pdf) for directions on how to obtain an ATP-ADP titration curve.
Conceptually, here is what we are doing to get the IC₅₀.
Plot the emission ratio of the 665 nm/615 nm RFU to the log of the concentration of the compound, log [inhibitor]. The ½ point on the curve as determined by the software of your choice is the assay EC₅₀.
- The emission ratio with no inhibitor present is about 0.27.
- The emission ratio at the EC₅₀ is about 0.56.
- The EC₅₀ is 35 nM.
Use the ATP-ADP titration curve to convert the emission ratios to percent of ATP converted.
- The percent conversion with no inhibitor present at an emission ratio of 0.27, black dashed line below, is 10% conversion of ATP to ADP.
- The percent conversion at the EC₅₀ with an emission ratio of 0.56, red dashed line, is about 3% conversion of ATP to ADP. This drop from 10% conversion to 3% conversion means that only 30% of the kinase activity is left, or about 70% inhibition. We are looking for 50% inhibition.
Note: If this seems confusing just think of it in arbitrary units. If the kinase activity dropped from 10 to 3, it is clear that the activity has gone down 70%. The confusion comes from different usages of the word percent.
- A 50% decrease in the activity of the kinase would represent a drop in the ATP conversion from 10% to 5%.
Note: Really low rates of ATP conversion are normal for this assay and desirable.
Use the ATP-ADP curve again, this time to find 5% conversion of ATP to ADP.
- 5% conversion corresponds to an emission ratio of 0.4.
Return to the plot of the emission ratio vs. log [inhibitor].
- An emission ratio of 0.4 yields an IC₅₀ of 12 nM.
Note: As discussed above, the EC₅₀, 35 nM is right-shifted compared to the true value of the IC₅₀, 12 nM.
Find additional tips, troubleshooting help, and resources within our Drug Discovery & Development Support Center.
As more inhibitor is present, less ADP is formed and more ADP Tracer stays bound to the antibody. Therefore, as the concentration of inhibitor increase, the signal increases.
In an assay in which the signal response is linear (or nearly linear) with respect to product formation, such as the LanthaScreen Eu Kinase Binding Assays, Z-LYTE Assays and others, the concentration of inhibitor that results in a 50% decrease in the assay signal, the EC₅₀, as determined by the emission ratio is equivalent to the IC₅₀ value for that inhibitor.
Since the response of the Adapta Universal Kinase Assay to ADP formation is not linear, the concentration of inhibitor required to cause a 50% decrease in the Emission Ratio, the EC₅₀, is NOT equivalent to the IC₅₀ value for the inhibitor.
To convert an inhibitor EC₅₀ value to an IC₅₀ value, use an ATP-ADP standard curve to correct for the non-linear response of the Adapta assay. ADP and ATP are provided in the kit. The curve is generated at constant total ATP plus ADP with known amounts of ATP and ADP.
Note: The substrate should be included in this titration.
Please refer to the Adapta Universal Kinase Assay User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/adapta_userguide_man.pdf) for directions on how to obtain an ATP-ADP titration curve.
Conceptually, here is what we are doing to get the IC₅₀.
Plot the emission ratio of the 665 nm/615 nm RFU to the log of the concentration of the compound, log [inhibitor]. The ½ point on the curve as determined by the software of your choice is the assay EC₅₀.
- The emission ratio with no inhibitor present is about 0.27.
- The emission ratio at the EC₅₀ is about 0.56.
- The EC₅₀ is 35 nM.
Use the ATP-ADP titration curve to convert the emission ratios to percent of ATP converted.
- The percent conversion with no inhibitor present at an emission ratio of 0.27, black dashed line below, is 10% conversion of ATP to ADP.
- The percent conversion at the EC₅₀ with an emission ratio of 0.56, red dashed line, is about 3% conversion of ATP to ADP. This drop from 10% conversion to 3% conversion means that only 30% of the kinase activity is left, or about 70% inhibition. We are looking for 50% inhibition.
Note: If this seems confusing just think of it in arbitrary units. If the kinase activity dropped from 10 to 3, it is clear that the activity has gone down 70%. The confusion comes from different usages of the word percent.
- A 50% decrease in the activity of the kinase would represent a drop in the ATP conversion from 10% to 5%.
Note: Really low rates of ATP conversion are normal for this assay and desirable.
Use the ATP-ADP curve again, this time to find 5% conversion of ATP to ADP.
- 5% conversion corresponds to an emission ratio of 0.4.
Return to the plot of the emission ratio vs. log [inhibitor].
- An emission ratio of 0.4 yields an IC₅₀ of 12 nM.
Note: As discussed above, the EC₅₀, 35 nM is right-shifted compared to the true value of the IC₅₀, 12 nM.
Find additional tips, troubleshooting help, and resources within our Drug Discovery & Development Support Center.