Product Image

Kinase Buffer S (5X)

Kinase Buffer S (5X)
Have Questions?
Catalog NumberQuantity
PV52134 mL
Catalog number PV5213
Price (USD)
244.00
Each
Quantity:
4 mL
Price (USD)
244.00
Each

Kinase buffer that has been specially optimized for use with LRRK2 and LRRK2 mutants.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chemical Name or MaterialKinase Buffer S
Includes5X Kinase Buffer S may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).
Concentration5X
For Use With (Equipment)LRRK2 and LRRK2 mutants
Physical FormLiquid
Quantity4 mL
Unit SizeEach